Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGXT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597036 Supplier Invitrogen™ Supplier No. PA597036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSVASLGGTP LYMDRQGIDI LYSGSQKALN APPGTSLISF SDKAKKKMYS RKTKPFSFYL DIKWLANFWG CDDQPRMYHH TIPVISLYSL RESLALIAEQ GLENSWRQHR EAAAYLHGRL QALGLQLFVK DPALRLPTVT TVAVPAGYDW RDIVSYVIDH FDIEIMGGLG PSTGKVLRIG LLGCNATREN VDRVTEALRA ALQHCPKKKL; Positive Samples: LO2, 293T; Cellular Location: Mitochondrion, Peroxisome.

This gene is expressed only in the liver and the encoded protein is localized mostly in the peroxisomes, where it is involved in glyoxylate detoxification. Mutations in this gene, some of which alter subcellular targetting, have been associated with type I primary hyperoxaluria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGXT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGXT
Gene Accession No. P21549
Gene Alias AGT; Agt1; Agxt; Agxt1; alanine-glyoxylate aminotransferase; Alanine--glyoxylate aminotransferase; alanine-glyoxylate aminotransferase 1; angiotensin receptor 2; hepatic peroxisomal alanine:glyoxylate aminotransferase; L-alanine: glyoxylate aminotransferase 1; PH1; serine:pyruvate aminotransferase; serine:pyruvate aminotransferase SPT; serine:pyruvate/alanine:glyoxylate aminotransferase; serine-pyruvate aminotransferase; Serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase, peroxisomal; serine--pyruvate aminotransferase; serine--pyruvate aminotransferase, mitochondrial; serine--pyruvate aminotransferase, peroxisomal; Spat; SPT; TLH6
Gene Symbols AGXT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 183-392 of human AGXT (NP_0000211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 189
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.