Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGXT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598871
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598871 Supplier Invitrogen™ Supplier No. PA598871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGXT2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by this gene is a class III pyridoxal-phosphate-dependent mitochondrial aminotransferase. It catalyzes the conversion of glyoxylate to glycine using L-alanine as the amino donor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGXT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Agxt2
Gene Accession No. Q9BYV1
Gene Alias (R)-3-amino-2-methylpropionate--pyruvate transaminase; AGT 2; Agt2; Agxt2; AI303810; AI663818; alanine-glyoxylate aminotransferase 2; alanine--glyoxylate aminotransferase 2; alanine--glyoxylate aminotransferase 2, mitochondrial; beta-ALAAT II; Beta-alanine-pyruvate aminotransferase; DAIBAT; D-AIBAT
Gene Symbols Agxt2
Host Species Rabbit
Immunogen Recombinant Human Alanine--glyoxylate aminotransferase 2, mitochondrial protein (196-328AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64902
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.