Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AHA1 Monoclonal Antibody (25F2.D9)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA527641
Description
1 μg/mL of MA5-27641 was sufficient for detection of Aha1 in 10 μg of rat tissue lysate by colorimetric immunoblot analysis using Goat anti-rat IgG:HRP as the secondary antibody. Detects approximately 38kDa. Can run up to 45kDa on SDS Page.
Acts as a co-chaperone of HSP90AA1 (PubMed:29127155). Activates the ATPase activity of HSP90AA1 leading to increase in its chaperone activity (PubMed:29127155). Competes with the inhibitory co-chaperone FNIP1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:27353360). Competes with the inhibitory co-chaperone TSC1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:29127155). [UniProt].
Specifications
AHA1 | |
Monoclonal | |
1 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and no preservative; pH 7.2 | |
O95433, Q8BK64 | |
Ahsa1 | |
Full-length mouse Aha1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
25F2.D9 | |
Unconjugated | |
Ahsa1 | |
Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W | |
Rat | |
Protein G | |
RUO | |
10598, 217737, 681996 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction