Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHA1 Monoclonal Antibody (4F3-B6)
GREENER_CHOICE

Catalog No. PIMA521096
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521096 Supplier Invitrogen™ Supplier No. MA521096
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAKWGEGDPR WIVEERANAT NVNNWHWTER DASNWSTDKL KTLFLAVQVQ NEEGKCEVTE VSKLDGEASI NNRKGKLIFF YEWSVKLNWT GTSKSGVQYK GHVEIPNLSG ENSVDEVEIS VSLAKDEPDT NLVALMKEEG VKLLREAMGI YISTLKTEFT QGMILPTMNG ESVDPVGQPA LKTEERKAKP APSKTQARPV GVKIPTCKIT LKETFLTSPE ELYRVFTTQE LVQAFTHAPA TLEADRGGKF HMVDGNVSGE FTDLVPEKHI VMKWRFKSWP EGHFATITLT FIDKNGETEL CMEGRGIPAP EEERTRQGWQ RYYFEGIKQT FGYGARLF.

Acts as a co-chaperone of HSP90AA1 (PubMed:29127155). Activates the ATPase activity of HSP90AA1 leading to increase in its chaperone activity (PubMed:29127155). Competes with the inhibitory co-chaperone FNIP1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:27353360). Competes with the inhibitory co-chaperone TSC1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:29127155). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHA1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4F3-B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Ahsa1
Gene Accession No. O95433
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W
Gene Symbols Ahsa1
Host Species Mouse
Immunogen AHSA1 (AAH00321, 1 a.a. ~ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10598
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.