Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114831 Supplier Invitrogen™ Supplier No. PA5114831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AHSA1.

Acts as a co-chaperone of HSP90AA1 (PubMed:29127155). Activates the ATPase activity of HSP90AA1 leading to increase in its chaperone activity (PubMed:29127155). Competes with the inhibitory co-chaperone FNIP1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:27353360). Competes with the inhibitory co-chaperone TSC1 for binding to HSP90AA1, thereby providing a reciprocal regulatory mechanism for chaperoning of client proteins (PubMed:29127155). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Ahsa1
Gene Accession No. O95433
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W
Gene Symbols Ahsa1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AHSA1(Accession O95433), corresponding to amino acid residues S84-A134.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10598
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.