Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHCTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556812 Supplier Invitrogen™ Supplier No. PA556812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEEARNLSFN ELYPSGTLKL QYNFDTIDQQ FCDLADNKDT AECDIAEVDG ELFVAQSNFT LILEGEEGEV EPGDFASSDV LPKAANTATE EKLVCSGEND Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.

Required for the assembly of a functional nuclear pore complex (NPC) on the surface of chromosomes as nuclei form at the end of mitosis. May initiate NPC assembly by binding to chromatin and recruiting the Nup107-160 subcomplex of the NPC. Also required for the localization of the Nup107-160 subcomplex of the NPC to the kinetochore during mitosis and for the completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHCTF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AHCTF1
Gene Accession No. Q8WYP5
Gene Alias 6230412P20Rik; Ahctf1; AT hook containing transcription factor 1; AT-hook containing transcription factor 1; AV011447; Elys; ELYS transcription factor like protein TMBS62; ELYS transcription factor-like protein TMBS62; embryonic large molecule derived from yolk sac; MST108; MSTP108; Protein ELYS; Protein MEL-28; Putative AT-hook-containing transcription factor 1; RGD1561291; TMBS62
Gene Symbols AHCTF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AHCTF1. Recombinant protein control fragment (Product #RP-94305).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.