Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AHDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140052
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RPPVLAKGDD PLPPRAARPV SQARCPTPVG DGSSSRRCWD NGRVNLRPVV QLIDIMKDLT RLSQDLQHSG VHLDCGGLRL SRPPAPPPGD LQYSFFS.

Specifications
AHDC1 | |
Polyclonal | |
Unconjugated | |
AHDC1 | |
AHDC1; AT hook, DNA binding motif, containing 1; AT-hook DNA binding motif containing 1; AT-hook DNA-binding motif-containing protein 1; MRD25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27245 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TGY3 | |
AHDC1 | |
Recombinant protein corresponding to Human AHDC1. Recombinant protein control fragment (Product #RP-108783). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction