Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ahlstrom-Munksjo Qualitative Filter Paper
Click to view available options
Filter Type:
Ashless
Hardened Ashless
Low Ash
Diameter (Metric):
21 mm
47 mm
55 mm
70 mm
90 mm
110 mm
125 mm
185 mm
240 mm
270 mm
320 mm
400 mm
425 mm
500 mm
Filtration Speed:
Fast Speed
Medium Speed
Slow Speed
Description
- For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
- High purity filters
- Consistent performance
- Available in a variety of sizes

Specifications
Specifications
Filter Type | Low Ash |
Format | Disc |
Filtration Speed | Fast Speed |
Type | Qualitative Filter Paper |
Diameter (Metric) | 110 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction