Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHNAK Polyclonal Antibody

AHNAK Polyclonal Antibody
Supplier: Thermo Scientific PA5106242
Description
AHNAK Polyclonal Antibody for Western Blot
Specifications
| AHNAK | |
| Polyclonal | |
| Unconjugated | |
| AHNAK | |
| AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66395, 79026 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q09666 | |
| AHNAK | |
| A synthesized peptide derived from human AHNAK(Accession Q09666), corresponding to amino acid residues E5758-L5808. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction