Learn More
Invitrogen™ AHNAK Recombinant Rabbit Monoclonal Antibody (PSH01-10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549610
Description
Sequence Similarities: 100% Mouse/Rat. Tissue Specificity: Low tissue specificity. Positive Control: Human lung carcinoma tissue, human brain tissue, mouse brain tissue, rat brain tissue, A431, HeLa, NIH/3T3. Subcellular Location: Nucleus. Predicted band size: 629 kDa.
AHNAK1 (Desmoyokin) is a large (700 kDa) scaffold protein that translocates to the plasma membrane after an increas of extracellular calcium level or upon proteinkinase C activation and regulates extracellular calcium influx mediated by L-type Ca2+ channels. AHNAK1 has been implicated in diverse signal transduction proceses affecting cell differentiation and proliferation. In response to calcium-dependent intercellular contacts AHNAK1 forms multimeric complexes in the plasma membrane, connected with actin and annexin 2/S100A10 assemblies and is thus involved in organization of the plasma membrane architecture. In epithelial cells, AHNAK1 is localized in cytoplasm or is membrane-associated, but in cells of nonepithelial origin AHNAK1 is predominantly nuclear; it has a weak DNA-binding activity and associates with the DNA-ligase IV-XRCC4 complex.
Specifications
AHNAK | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q09666 | |
AHNAK | |
Recombinant protein within human AHNAK aa 451-900/5,890. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
PSH01-10 | |
Unconjugated | |
AHNAK | |
AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118 | |
Rabbit | |
Protein A | |
RUO | |
191572, 66395, 79026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.