Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHNAK2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen AHNAK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1576620
SDP
View Documents
Novus Biologicals
NBP15766120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP157661
SDP
View Documents
Novus Biologicals
NBP157661
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

AHNAK2 Polyclonal specifically detects AHNAK2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AHNAK2
Polyclonal
Rabbit
AHNAK nucleoprotein 2, C14orf78, chromosome 14 open reading frame 78, KIAA2019, protein AHNAK2
AHNAK2
Primary
Western Blot
Unconjugated
RUO
113146
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.