Learn More
Description
This gene encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450.
- Aryl hydrocarbon receptor
- Transfected cell line: 293T
- Plasmid: pCMV-AHR full-length
- Host: Human
- Theoretical molecular weight: 96.1kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Cell Line | 293T |
| Description | AHR 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_001621.3 |
| Molecular Weight (g/mol) | 96.1 |
| pH Range | 6.8 |
| Plasmid | pCMV-AHR full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-AHR antibody (H00000196-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.