Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHR Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002289F
Description
AHR Polyclonal specifically detects AHR in Human, Mouse, Rat, Guinea Pig samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
AHR | |
Polyclonal | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
P35869 | |
AHR | |
Bacterially expressed human Aryl hydrocarbon Receptor (C-terminus). [UniProt# P35869] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse and Guinea Pig. Rat reactivity reported in scientific literature (PMID: 23887904). | |
Store at 4C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
FITC | |
PBS with 0.05% Sodium Azide | |
Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
Rabbit | |
96 kDa | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
196 | |
Human, Mouse, Rat, Guinea Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction