Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AhR Mouse anti-Human, Mouse, Alexa Fluor 488, Clone: T49-550, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565788
Description
AhR Monoclonal antibody specifically detects AhR in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
AhR | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |
Flow Cytometry | |
T49-550 | |
Alexa Fluor 488 | |
AHR; Aryl hydrocarbon R; Ah-Receptor; Aromatic hydrocarbon receptor; bHLHe7 | |
Human AhR Recombinant Protein | |
RUO | |
Human, Mouse | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction