Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHR Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002289AF350
Description
AHR Polyclonal specifically detects AHR in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
AHR | |
Polyclonal | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 | |
Rabbit | |
Affinity Purified | |
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Hypoxia, Innate Immunity, Transcription Factors and Regulators | |
196.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
AHR | |
Bacterially expressed human Aryl hydrocarbon Receptor (C-terminus). [UniProtNo. P35869] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Guinea Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction