Learn More
Invitrogen™ AHRR Polyclonal Antibody

Description
The antibody detects endogenous levels of total AHRR protein.
Specifications
Specifications
| Antigen | AHRR |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | AHRR |
| Gene Accession No. | A9YTQ3, Q3U1U7 |
| Gene Alias | AHH; AHHR; ahR repressor; AHRR; aryl hydrocarbon hydroxylase regulator; aryl hydrocarbon receptor repressor; aryl-hydrocarbon receptor repressor; BHLHE77; Class E basic helix-loop-helix protein 77; dioxin receptor repressor; KIAA1234; mKIAA1234 |
| Gene Symbols | AHRR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.