Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHRR Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$220.00 - $536.00
Specifications
| Antigen | AHRR |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5122263
|
Invitrogen™
PA5122263 |
20 μL |
Each for $220.00
|
|
|||||
PIPA5122262
![]() |
Invitrogen™
PA5122262 |
100 μL |
Each for $536.00
|
|
|||||
Description
The protein encoded by this gene participates in the aryl hydrocarbon receptor (AhR) signaling cascade, which mediates dioxin toxicity, and is involved in regulation of cell growth and differentiation. It functions as a feedback modulator by repressing AhR-dependent gene expression. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| AHRR | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A9YTQ3 | |
| 57491 | |
| Met1-Pro182 (Accession A9YTQ3), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| AHRR |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| AHH; AHHR; ahR repressor; AHRR; aryl hydrocarbon hydroxylase regulator; aryl hydrocarbon receptor repressor; aryl-hydrocarbon receptor repressor; BHLHE77; Class E basic helix-loop-helix protein 77; dioxin receptor repressor; KIAA1234; mKIAA1234 | |
| AHRR | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title