Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHRR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5107055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107055 Supplier Thermo Scientific Supplier No. PA5107055

Rabbit Polyclonal Antibody

Sequence homology: Mouse: 100%; Rat: 93%

The protein encoded by this gene participates in the aryl hydrocarbon receptor (AhR) signaling cascade, which mediates dioxin toxicity, and is involved in regulation of cell growth and differentiation. It functions as a feedback modulator by repressing AhR-dependent gene expression. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHRR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene AHRR
Gene Accession No. Q3U1U7
Gene Alias AHH; AHHR; ahR repressor; AHRR; aryl hydrocarbon hydroxylase regulator; aryl hydrocarbon receptor repressor; aryl-hydrocarbon receptor repressor; BHLHE77; Class E basic helix-loop-helix protein 77; dioxin receptor repressor; KIAA1234; mKIAA1234
Gene Symbols AHRR
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: CLRGGPDLLD PKGTSGDREE EDQKHILRRS PGAWGQREMH KYSYGLETPV
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11624
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.