Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHSA1 Antibody (4H9-D7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212891R
Description
AHSA1 Monoclonal antibody specifically detects AHSA1 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
AHSA1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
4H9-D7 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
Human Aha1, his tagged | |
0.1 mL | |
Signal Transduction | |
10598 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction