Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHSA1 Antibody (SR1727), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322144100UL
Description
AHSA1 Monoclonal antibody specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
AHSA1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1727 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
A synthesized peptide derived from human AHSA1 (Uniprot #: O95433) | |
100 μg | |
Signal Transduction | |
10598 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction