Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AHSA1 Mouse anti-Human, Clone: 7A6, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | AHSA1 |
---|---|
Clone | 7A6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1006, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25939350
![]() |
Novus Biologicals
NBP25939350UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259393X
![]() |
Novus Biologicals
NBP259393100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
AHSA1 Monoclonal specifically detects AHSA1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AHSA1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
AHSA1 | |
IgG1 κ | |
Protein A purified |
7A6 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1006, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
10598 | |
Recombinant human COX5A (42-150aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title