Learn More
AHSA1 Mouse anti-Human, Clone: OTI1D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808924

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
AHA1 is a co-chaperone that stimulates HSP90 ATPase activity, and may affect a step in the endoplasmic reticulum to Golgi trafficking.Specifications
AHSA1 | |
Monoclonal | |
Unconjugated | |
AHSA1 | |
AHA1, C14orf3, hAha1, p38 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10598 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95433 | |
AHSA1 | |
Human recombit protein fragment corresponding to amino acids 1-203 of human AHSA1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.