Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHSA2 Monoclonal Antibody (3G11)
GREENER_CHOICE

Catalog No. PIMA524516
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524516 Supplier Invitrogen™ Supplier No. MA524516
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MILPTKAMAT QELTVKRKLS GNTLQVQASS PVALGVRIPT VALHMMELFD TTVEQLYSIF TVKELTNKKI IMKWRCGNWP EEHYAMVALN FVPTLGQTEL QLKEFLSICK EENMKFCWQK QHFEEIKGSL QLTPLNG.

Co-chaperone that stimulates HSP90 ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSA2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G11
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Ahsa2
Gene Accession No. Q719I0
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 2; activator of heat shock protein ATPase homolog 2; AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast); AHA1, activator of heat shock protein ATPase 2; AHA1, activator of heat shock protein ATPase homolog 2; Ahsa2; Hch1
Gene Symbols Ahsa2
Host Species Mouse
Immunogen AHSA2 (NP_689605.1, 1 a.a. ~ 137 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130872
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.