Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHSA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113017
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113017 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113017 Supplier Invitrogen™ Supplier No. PA5113017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AHSA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Co-chaperone that stimulates HSP90 ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ahsa2
Gene Accession No. Q719I0, Q8N9S3
Gene Alias Activator of 90 kDa heat shock protein ATPase homolog 2; Activator of heat shock protein ATPase homolog 2; AHA1, Activator of heat shock 90kDa protein ATPase homolog 2 (yeast); AHA1, Activator of heat shock protein ATPase 2; AHA1, Activator of heat shock protein ATPase homolog 2; Ahsa2; Hch1
Gene Symbols Ahsa2
Host Species Rabbit
Immunogen Recombinant Human Putative activator of 90 kDa heat shock protein ATPase homolog 2 protein (1-137AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130872, 268390, 305577
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.