Learn More
AHSG Mouse anti-Human, Clone: OTI2H2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504670

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.Specifications
AHSG | |
Monoclonal | |
Unconjugated | |
AHSG | |
A2HS, AHS, APMR1, FETUA, HSGA | |
Mouse | |
Affinity Chromatography | |
RUO | |
197 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2H2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02765 | |
AHSG | |
Full length human recombit protein of human AHSG produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.