Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AHSG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985012
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5012 Supplier Bioss Supplier No. BS2922R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.

Specifications

Antigen AHSG
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AHSG
Gene Accession No. P02765
Gene Alias 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; Ahsg; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Asialofetuin; Ba-alpha-2-glycoprotein; BOS_906; BSP; Countertrypin; FETUA; fetuin A; FetuinA; Fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743
Gene Symbols AHSG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Alpha-2-HS-glycoprotein chain A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.