Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AHSP Polyclonal Antibody

Catalog No. PIPA5122260
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122260 20 μL
PIPA5118943 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122260 Supplier Invitrogen™ Supplier No. PA5122260
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AHSP Polyclonal Antibody for IHC (P), Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AHSP
Gene Accession No. Q9CY02
Gene Alias AHSP; alpha hemoglobin stabilising protein; alpha hemoglobin stabilizing protein; alpha-hemoglobin-stabilizing protein; Edrf; Eraf; erythroid associated factor; erythroid differentiation associated factor; erythroid differentiation-related factor; Erythroid-associated factor
Gene Symbols AHSP
Host Species Rabbit
Immunogen Recombinant protein Alpha-Hemoglobin Stabilizing Protein. The antigen corresponds to amino acid range 2-102 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170812
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.