Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AHSP Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | AHSP |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118942
|
Invitrogen™
PA5118942 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122261
|
Invitrogen™
PA5122261 |
20 μL |
Each for $209.50
|
|
|||||
Description
AHSP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
AHSP | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse, Rat | |
AHSP | |
AHSP; alpha hemoglobin stabilising protein; alpha hemoglobin stabilizing protein; alpha-hemoglobin-stabilizing protein; Edrf; Eraf; erythroid associated factor; erythroid differentiation associated factor; erythroid differentiation-related factor; Erythroid-associated factor | |
AHSP | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9CY02 | |
170812, 293522 | |
Recombinant protein Alpha-Hemoglobin Stabilizing Protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title