Learn More
AIB-1 Mouse, Unlabeled, Clone: 34, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611105
Description
Signal transduction via nuclear hormone receptors is important for cell growth and differentiation, development, and homeostasis. Nuclear hormone receptors are ligand-activated transcription factors that modulate target gene expression. These ligand/receptor complexes also interact with transcriptional coactivators which enhance ligand-dependent transcription. Various classes of coactivators have been identified, including SRC-1 and its related proteins, such as TIF-2/GRIP-1, RIP140, AIB-1, and TIF-1α and -1β. AIB-1 (Amplified In Breast cancer-1) is also known as pCIP, RAC3, and TRAM-1. It interacts with estrogen receptor (ER) and is overexpressed in breast cancer biopsies and several breast and ovarian cancer cell lines. Similar to SRC-1 and TIF2, AIB-1 contains a basic helix-loop-helix (bHLH) domain followed by a PAS (Per/Arnt/Sim) region, serine and threonine rich regions, and a charged cluster. In addition, AIB-1 contains three copies of the conserved LXXLL motif which is critical to its interaction with the nuclear receptor. Thus, AIB-1 is a coactivator of nuclear receptors that may participate in the development of steroid-dependent cancers.

Specifications
AIB-1 | |
Monoclonal | |
250μg/mL | |
Amplified In Breast cancer-1; pCIP; RAC3; TRAM-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
34 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human AIB-1 aa. 376-389 | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.