Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRC3 Monoclonal Antibody (5H3)
GREENER_CHOICE

Catalog No. PIMA515898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515898 Supplier Invitrogen™ Supplier No. MA515898
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15898 targets AIB1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15898 immunogen is purified recombinant fragment of human AIB1 expressed in E. Coli. . MA5-15898 detects AIB1 which has a predicted molecular weight of approximately 155kDa.

The poly Q polymorphism in AIB1 (amplified in breast cancer) gene is usually assessed by fragment length analysis which does not reveal the actual sequence variation. The purpose of this study is to investigate the sequence variation of poly Q encoding region in breast cancer cell lines at single molecule level, and to determine if the sequence variation is related to AIB1 gene amplification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRC3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5H3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene NCOA3
Gene Accession No. Q9Y6Q9
Gene Alias ACTR; AIB1; AIB-1; Amplified in breast cancer 1 protein; bHLHe42; CAGH16; CBP-interacting protein; class E basic helix-loop-helix protein 42; CTG26; KAT13B; NCOA3; NCoA-3; Nuclear receptor coactivator 3; pCIP; RAC3; RAC-3; receptor-associated coactivator 3; SRC3; SRC-3; Steroid receptor coactivator protein 3; thyroid hormone receptor activator molecule 1; TNRC14; TNRC16; TRAM1; TRAM-1
Gene Symbols NCOA3
Host Species Mouse
Immunogen Purified recombinant fragment of human AIB1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.