Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIB3 Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 39836

Description
Nuclear receptor coactivator that directly binds nuclear receptors and stimulates the transcriptional activities in a hormone-dependent fashion. Coactivates expression in an agonist- and AF2-dependent manner. Involved in the coactivation of different nuclear receptors, such as for steroids (GR and ERs), retinoids (RARs and RXRs), thyroid hormone (TRs), vitamin D3 (VDR) and prostanoids (PPARs). Probably functions as a general coactivator, rather than just a nuclear receptor coactivator. May also be involved in the coactivation of the NF-kappa-B pathway. May coactivate expression via a remodeling of chromatin and its interaction with histone acetyltransferase proteins.Specifications
AIB3 | |
Polyclonal | |
Unconjugated | |
NCOA6 | |
Activating signal cointegrator 2; activating signal cointegrator-2; AIB3; Amplified in breast cancer protein 3; amplified in breast cancer-3 protein; ASC2; ASC-2; Cancer-amplified transcriptional coactivator ASC-2; KIAA0181; mKIAA0181; NCOA6; Ncoa7; NRC; NRC RAP250; Nuclear receptor coactivator 6; Nuclear receptor coactivator RAP250; nuclear receptor-activating protein 250; Nuclear receptor-activating protein, 250 kDa; peroxisome proliferator-activated receptor interacting protein; peroxisome proliferator-activated receptor interacting protein,PRIP; Peroxisome proliferator-activated receptor-interacting protein; PPAR interacting protein PRIP; PPAR-interacting protein; PRIP; RAP250; thyroid hormone receptor binding protein; Thyroid hormone receptor-binding protein; TRBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23054, 56406 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Lot-Specific | |
70mM tris HCl with 1% BSA, 105mM NaCl, 30% glycerol, 70mM glycine and 0.035% sodium azide; pH 8 | |
Q14686, Q9JL19 | |
NCOA6 | |
This AIB3 antibody was raised against a recombinant protein corresponding to amino acids 22-146 of mouse AIB3. | |
10 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction