Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AIDA
GREENER_CHOICESDP

Catalog No. 090450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-045-0 50 mg
09-041-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-045-0 Supplier R&D Systems™ Supplier No. 0904/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective group I mGlu antagonist

A relatively potent and selective antagonist of group I metabotropic glutamate receptors (mGlu1a), having no effect on group II (mGlu2) or group III (mGlu4) receptors expressed individually in baby hamster kidney cells. Has no effect on ionotropic glutamate receptors. Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 168560-79-0
Molecular Formula C11H10NO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 221.21
Purity >98%
Molecular Weight (g/mol) 220.21
Chemical Name or Material AIDA
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.