Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIDA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555734 Supplier Invitrogen™ Supplier No. PA555734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEENLEFEED EEEGGAGAGS PDSFPARVPG TLLPRLPSEP GMTLLTIRIE KIGLKDAGQC IDPYITVS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

AIDA acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIDA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AIDA
Gene Accession No. Q96BJ3
Gene Alias 2610208M17Rik; AIDA; Axin interaction partner and dorsalization antagonist; axin interactor, dorsalization associated; axin interactor, dorsalization-associated protein; BC004835; C1orf80
Gene Symbols AIDA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AIDA. Recombinant protein control fragment (Product #RP-94510).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.