Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF-1/Iba1 Antibody (031), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290271MFV450
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
AIF-1/Iba1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
031 | |
ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
Rabbit immunized with purified, recombinant Human AIF-1/Iba1 (Uniprot#: P55008-1, Met1-Pro147) | |
0.1 mL | |
Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
199 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction