Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF-1/Iba1 Antibody (603102) [PerCP], Novus Biologicals™
SDP

Catalog No. FAB7308C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7308C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB7308C Supplier Novus Biologicals Supplier No. FAB7308C
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen AIF-1/Iba1
Applications Immunohistochemistry
Classification Monoclonal
Clone 603102
Conjugate PerCP
Formulation PBS
Gene Alias AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Host Species Mouse
Immunogen E. coli-derived recombinant human AIF-1/Iba1, Met1-Pro147, Accession # P55008
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 199
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.