Learn More
AIF-1/Iba1 Antibody (rAIF1/1909), Janelia Fluor™ 549, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP275760JF549
Catalog No. NB008954
Only null left
Add to Cart
Description
Description
AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
AIF-1/Iba1
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
AIF1
Purified recombinant human AIF-1/Iba1 protein (around aa 1-146) (exact sequence is proprietary) (Uniprot: P55008)
0.1 mL
Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
AIF1 is a cytoplasmic, calcium-binding protein that is thought to play a role in macrophage activation and function. AIF1, containing two EF domains, is induced by cytokines and Interferons. In an unstimulated state, AIF1 colocalizes with actin, and upon stimulation, translocates to lamellipodia. It is also a marker of human microglia and is expressed by macrophages in injured skeletal muscle. The gene encoding AIF1 resides in the tumor necrosis factor (TNF) cluster of genes, located in the region represented by the human major histocompatibility complex (MHC).
Store at 4°C in the dark.
Immunohistochemistry, Immunohistochemistry (Paraffin)
rAIF1/1909
Immunohistochemistry, Immunohistochemistry-Paraffin
AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Mouse
Protein A or G purified
RUO
Primary
Human
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.