Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF/AIFM1 Mouse anti-Human, Clone: 2.20E+10, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259432100UL
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
AIF/AIFM1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1039, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
AIFM1 apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, PDCD8 | |
Mouse | |
Protein A purified | |
RUO | |
9131 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
22000000000 | |
Unconjugated | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
AIFM1 | |
Recombinant human AIFM1 (98-609aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction