Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF Antibody (22E9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259432F
This item is not returnable.
View return policy
Description
AIF Monoclonal antibody specifically detects AIF in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
AIF | |
Monoclonal | |
FITC | |
AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
Recombinant human AIF (98-609aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
9131 | |
Store at 4°C in the dark. | |
IgG2a κ |
ELISA, Immunofluorescence | |
22000000000 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction