Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF Antibody (3C11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2506060.025ML
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot.Specifications
AIF | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100 - 1:2000 | |
AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
Mouse | |
Protein A purified | |
RUO | |
9131 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG1 |
Western Blot | |
3C11 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
AIFM1 | |
AIF-His fusion protein | |
0.025 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction