Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF Antibody (3C11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250606G
Description
AIF Monoclonal specifically detects AIF in Human samples. It is validated for Western Blot.Specifications
AIF | |
Monoclonal | |
DyLight 488 | |
50 mM sodium borate with 0.05% sodium azide | |
AIFM1 | |
AIF-His fusion protein | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
3C11 | |
Western Blot | |
AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
Mouse | |
Protein A purified | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
9131.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction