Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $390.00
Specifications
Antigen | AIF |
---|---|
Dilution | Western Blot 1:1000-1:2000∼, Simple Western 1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227177SS
![]() |
Novus Biologicals
NBP227177SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP227177
![]() |
Novus Biologicals
NBP227177 |
0.05 mL |
Each for $390.00
|
|
|||||
Description
AIF Polyclonal specifically detects AIF in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
AIF | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Mitochondrial Markers, Tumor Suppressors | |
50 ul neat whole antisera with 0.05% Sodium Azide | |
AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
AIFM1 | |
IgG | |
Unpurified |
Western Blot 1:1000-1:2000∼, Simple Western 1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat, Bovine, Canine | |
O95831 | |
9131 | |
(aa 151-170); human | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title