Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF1 Rabbit anti-Human, Unconjugated, Clone: rAIF1/1909, Abnova™

Catalog No. 89337903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-903 Supplier Abnova Corporation Supplier No. RAB00478
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

AIF1 Monoclonal antibody specifically detects AIF1 in Human samples. It is validated for IHC-P.

Specifications

Antigen AIF1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rAIF1/1909
Conjugate Unconjugated
Description allograft inflammatory factor 1
Gene AIF1
Gene Alias AIF-1, IBA1, IRT-1
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acids 1-146 of human AIF1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 199
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.