Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIF1L Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299233MFV450
Description
AIF1L Polyclonal antibody specifically detects AIF1L in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
AIF1L | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
allograft inflammatory factor 1-like, C9orf58, chromosome 9 open reading frame 58, IBA2FLJ12783, ionized calcium binding adapter molecule 2, Ionized calcium-binding adapter molecule 2, MGC29466 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human AIF1L. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
83543 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction