Learn More
AIF1L Polyclonal Antibody

Description
AIF1L Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | AIF1L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | AIF1L |
| Gene Accession No. | Q9BQI0 |
| Gene Alias | AIF1L; allograft inflammatory factor 1 like; allograft inflammatory factor 1-like; C9orf58; IBA2; ionized calcium binding adapter molecule 2; ionized calcium-binding adapter molecule 2; UNQ672/PRO1306 |
| Gene Symbols | AIF1L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.