Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIFM3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176378
Description
AIFM3 Polyclonal specifically detects AIFM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
AIFM3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
AIFLFLJ30473, apoptosis-inducing factor 3, apoptosis-inducing factor like, apoptosis-inducing factor, mitochondrion-associated, 3, Apoptosis-inducing factor-like protein, FLJ45137 | |
Rabbit | |
Affinity Purified | |
RUO | |
150209 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
Q96NN9 | |
AIFM3 | |
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human AIFM3. The immunogen is located within amino acids 210 - 260 of AIFM3. Amino Acid Squence: DRHLPYDRPKLSKSLD | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction