Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIFM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597129 Supplier Invitrogen™ Supplier No. PA597129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGGCFSKPKP VELKIEVVLP EKERGKEELS ASGKGSPRAY QGNGTARHFH TEERLSTPHP YPSPQDCVEA AVCHVKDLEN GQMREVELGW GKVLLVKDNG EFHALGHKCP HYGAPLVKGV LSRGRVRCPW HGACFNISTG DLEDFPGLDS LHKFQVKIEK EKVYVRASKQ ALQLQRRTKV MAKCISPSAG YSSSTNVLIV; Positive Samples: THP-1, HeLa; Cellular Location: Mitochondrion.

Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Recently a protein similar to the apoptosis-inducing factor (AIF) was cloned and designated AIFL (also known as AIFM3). AIFM3 is expressed ubiquitously and is predominantly localized to the inner membranes of mitochondria. Unlike AIF, AIFM3 does not translocate to the nucleus upon induction of apoptosis. However, overexpression of AIFM3, like AIF, induced cytochrome c release from the mitochondria, cleavage of caspase 3, and ultimately apoptosis, indicating AIFM3 induces apoptosis through caspase activation. Multiple isoforms of AIFM3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIFM3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AIFM3
Gene Accession No. Q3TY86, Q96NN9
Gene Alias 2810401C16Rik; AI840249; Aifl; AIFM3; apoptosis inducing factor, mitochondria associated 3; apoptosis-inducing factor 3; apoptosis-inducing factor like; apoptosis-inducing factor, mitochondrion-associated 3; apoptosis-inducing factor, mitochondrion-associated, 3; apoptosis-inducing factor-like protein; RGD1306028
Gene Symbols AIFM3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AIFM3 (NP_0010180701).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150209, 303786, 72168
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.