Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AIG1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | AIG1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AIG1 Polyclonal specifically detects AIG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AIG1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
51390 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human AIG1. The immunogen is located within amino acids 50 - 100 of AIG1. Amino Acid Squence: LTRGSGNQEQERQLKKL | |
Primary |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
androgen-induced 1, androgen-induced gene 1 protein, C-terminus truncated, DKFZp686F03136 | |
AIG1 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title