Learn More
Invitrogen™ AIG1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total AIG1.
Specifications
Specifications
| Antigen | AIG1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | AIG1 |
| Gene Accession No. | Q9NVV5 |
| Gene Alias | 1500031O19Rik; AIG1; AIG-1; androgen induced 1; androgen-induced 1; androgen-induced gene 1 protein; AV064870; AW413422; CGI-103; dJ95L4.1; FAHFA hydrolase AIG1; Fatty acid esters of hydroxy fatty acids hydrolase AIG1; RGD1562920 |
| Gene Symbols | AIG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.