Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729475 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9475 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9475 Supplier Proteintech Group Inc Supplier No. 144681AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Androgen-induced 1 (AIG1) is a novel Pirh2-interacting protein, which was initially cloned from human dermal papilla cells and is evolutionally conserved from drosophila through to humans. It is a membrane protein and belongs to the protein family containing FAR-17a domain. AIG1 may play a role in androgen-regulated growth of hair follicles. AIG1 could serve as a new biomarker for the diagnosis and prognostic evaluation of HCCs and may be a novel candidate tumor suppressor.

Specifications

Antigen AIG1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AIG1
Gene Accession No. Q9D8B1, Q9NVV5
Gene Alias AIG 1, AIG1, androgen induced 1
Gene Symbols AIG1
Host Species Rabbit
Immunogen AIG1 Fusion Protein Ag5836
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292486, 51390, 66253
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.