Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AIM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584699 Supplier Invitrogen™ Supplier No. PA584699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCSGREATLQ DCPSGPWGKN TCNHDEDTWV ECEDPFDLRL VGGDNLCSGR LEVLHKGVWG SVCDDNWGEK E Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 72%.

CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIM
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cd5l
Gene Accession No. O43866
Gene Alias 1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; CD5L; CT2; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; igM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; Spalpha; Sp-alpha; UNQ203/PRO229
Gene Symbols Cd5l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CD5L. Recombinant protein control fragment (Product #RP-103606).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.